Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 323/A3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453201

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Quantity 0.1mg
Isotype IgG1, κ
Antigen EpCAM/TROP1
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Gene Accession No. P16422
Gene ID (Entrez) 4072
Test Specificity Does not react to Rat.
Cross Reactivity Human
Immunogen MCF-7 human breast cancer cells
Conjugate Unlabeled
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Clone 323/A3
Primary or Secondary Primary