Please login to your online account to display your discounted pricing

Mouse anti-EMMPRIN/CD147, Clone: CB43, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen EMMPRIN/CD147
Clone CB43
Immunogen Native human leucocytes
Gene Accession No. P35613
Gene ID (Entrez) 682
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23446801

 
novus biologicals canada
NBP23446801ML
0.1mL Each for $139.00

NBP23446805

 
novus biologicals canada
NBP23446805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen EMMPRIN/CD147
Clone CB43
Immunogen Native human leucocytes
Gene Accession No. P35613
Gene ID (Entrez) 682
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)