Please login to your online account to display your discounted pricing

Mouse anti-EMMPRIN/CD147, Clone: CB43, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446805

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Monoclonal or Polyclonal Monoclonal
Immunogen Native human leucocytes
Cross Reactivity Human
Host Species Mouse
Conjugate Unlabeled
Antigen EMMPRIN/CD147
Purification Method Concentrated
Quantity 0.5mL
Gene ID (Entrez) 682
Gene Accession No. P35613
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Isotype IgM
Regulatory Status RUO
Clone CB43
Primary or Secondary Primary