Please login to your online account to display your discounted pricing

Mouse anti-EMMPRIN/CD147, Clone: CB43, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446801

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P35613
Host Species Mouse
Immunogen Native human leucocytes
Isotype IgM
Purification Method Concentrated
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 682
Cross Reactivity Human
Antigen EMMPRIN/CD147
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone CB43
Conjugate Unlabeled