missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-EMMPRIN/CD147, Clone: 8D6, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen EMMPRIN/CD147
Clone 8D6
Concentration 1mg/mL
Immunogen Human T cell line Molt 13
Gene Accession No. P35613
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23454301 Novus Biologicals Canada
NBP23454301MG
0.1mg Each for $384.00
NBP23454302 Novus Biologicals Canada
NBP23454302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

EMMPRIN/CD147
1mg/mL
P35613
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
8D6
Human T cell line Molt 13
682
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot