Please login to your online account to display your discounted pricing

Mouse anti-EMMPRIN/CD147, Clone: 8D6, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Immunogen Human T cell line Molt 13
Monoclonal or Polyclonal Monoclonal
Antigen EMMPRIN/CD147
Cross Reactivity Human
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone 8D6
Host Species Mouse
Isotype IgG1
Primary or Secondary Primary
Gene ID (Entrez) 682
Gene Accession No. P35613
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Reconstitution Protein A purified
Purification Method Purified