Please login to your online account to display your discounted pricing

Mouse anti-EGF R/ErbB1, Clone: H9B4, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen EGF R/ErbB1
Clone H9B4
Concentration 0.2mg/mL
Immunogen Purified EGFR from A431 cells.
Gene Accession No. P00533
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23424301

 
novus biologicals canada
NBP23424301ML
0.1mL Each for $139.00

NBP23424305

 
novus biologicals canada
NBP23424305ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen EGF R/ErbB1
Clone H9B4
Concentration 0.2mg/mL
Immunogen Purified EGFR from A431 cells.
Gene Accession No. P00533
Gene ID (Entrez) 1956
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunoprecipitation
Applications Western Blotting