Please login to your online account to display your discounted pricing

Mouse anti-EGF R/ErbB1, Clone: H9B4, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23424305ML

 View all options for this product


For Research Use Only

Catalog No. NBP23424305


Description & Specifications


Antigen EGF R/ErbB1
Applications ELISA
Applications Immunoprecipitation
Applications Western Blotting
Clone H9B4
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P00533
Host Species Mouse
Immunogen Purified EGFR from A431 cells.
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1956
Test Specificity Does not react to Mouse or Rat.