Please login to your online account to display your discounted pricing

Mouse anti-EGF R/ErbB1, Clone: H9B4, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462801

  • / Each
 

Description & Specifications

Specifications

Antigen EGF R/ErbB1
Applications ELISA
Applications Immunoprecipitation
Applications Western Blot
Clone H9B4
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P00533
Host Species Murine
Immunogen Purified EGFR from A431 cells.
Isotype IgG1
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1956
Test Specificity Does not react to Mouse or Rat.
Reconstitution Protein A purified
Target Species Human