Please login to your online account to display your discounted pricing

Mouse anti-EGF R/ErbB1, Clone: B1D8, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23455301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23455301

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Neutralization
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen EGF R/ErbB1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P00533
Immunogen Microsomes from A431 cells
Isotype IgG2a, κ
Quantity 0.1mg
Regulatory Status RUO
Gene ID (Entrez) 1956
Conjugate Unlabeled
Host Species Mouse
Purification Method Purified
Reconstitution Protein A purified
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Does not react to Mouse or Rat.
Clone B1D8