Please login to your online account to display your discounted pricing

Mouse anti-EEA1, Clone: 5G3.2C2, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen EEA1
Clone 5G3.2C2
Concentration 1.0mg/mL
Immunogen Partial recombinant human EEA1 protein (between amino acids 50-300) [UniProt Q15075]
Gene Accession No. Q15075
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236569

 
novus biologicals canada
NBP236569
0.1mg Each for $259.02

NBP236569SS

 
novus biologicals canada
NBP236569SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen EEA1
Clone 5G3.2C2
Concentration 1.0mg/mL
Immunogen Partial recombinant human EEA1 protein (between amino acids 50-300) [UniProt Q15075]
Gene Accession No. Q15075
Gene ID (Entrez) 8411
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow (Intracellular)
Applications Flow Cytometry (Intracellular Staining)
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot