Please login to your online account to display your discounted pricing

Mouse anti-ECM-1/Secretory Component P85, Clone: SPM217, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen ECM-1/Secretory Component P85
Clone SPM217
Concentration 0.2mg/mL
Immunogen Secretory Component protein isolated from human colostrum
Gene Accession No. Q16610
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23286901

 
novus biologicals canada
NBP23286901ML
0.1mL Each for $139.00

NBP23286910

 
novus biologicals canada
NBP23286910ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen ECM-1/Secretory Component P85
Clone SPM217
Concentration 0.2mg/mL
Immunogen Secretory Component protein isolated from human colostrum
Gene Accession No. Q16610
Gene ID (Entrez) 1893
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting