Please login to your online account to display your discounted pricing

Mouse anti-ECM-1/Secretory Component P85, Clone: SPM217, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen ECM-1/Secretory Component P85
Clone SPM217
Concentration 1mg/mL
Immunogen Secretory Component protein isolated from human colostrum
Gene Accession No. Q16610
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23474701

 
novus biologicals canada
NBP23474701MG
0.1mg Each for $384.00

NBP23474702

 
novus biologicals canada
NBP23474702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen ECM-1/Secretory Component P85
Clone SPM217
Concentration 1mg/mL
Immunogen Secretory Component protein isolated from human colostrum
Gene Accession No. Q16610
Gene ID (Entrez) 1893
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot