Please login to your online account to display your discounted pricing

Mouse anti-ECM-1/Secretory Component P85, Clone: SPM217, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474702

  • / Each
 

Description & Specifications

Specifications

Antigen ECM-1/Secretory Component P85
Conjugate Unlabeled
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component.
Host Species Mouse
Gene ID (Entrez) 1893
Regulatory Status RUO
Clone SPM217
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Immunogen Secretory Component protein isolated from human colostrum
Gene Accession No. Q16610
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Reconstitution Protein A purified
Isotype IgG1, κ
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.