Please login to your online account to display your discounted pricing

Mouse anti-ECM-1/Secretory Component P85, Clone: SPM217, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474701

  • / Each
 

Description & Specifications

Specifications

Antigen ECM-1/Secretory Component P85
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM217
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q16610
Host Species Murine
Immunogen Secretory Component protein isolated from human colostrum
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1893
Test Specificity This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component.
Reconstitution Protein A purified
Target Species Human
Target Species Rat