Please login to your online account to display your discounted pricing

Mouse anti-EBV Early Antigens, Clone: 1108-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433305

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Antigen EBV Early Antigens
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Epstein-Barr virus
Reconstitution Protein A purified
Clone 1108-1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Virus