Please login to your online account to display your discounted pricing

Mouse anti-EBV Early Antigens, Clone: 1108-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen EBV Early Antigens
Clone 1108-1
Concentration 1mg/mL
Immunogen Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23451602

 
novus biologicals canada
NBP23451602MG
0.2mg Each for $527.00

NBP23451601

 
novus biologicals canada
NBP23451601MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen EBV Early Antigens
Clone 1108-1
Concentration 1mg/mL
Immunogen Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Virus
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting