Please login to your online account to display your discounted pricing

Mouse anti-EBV Early Antigens, Clone: 1108-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451602

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
Isotype IgG1, κ
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.2mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Epstein-Barr virus
Clone 1108-1
Concentration 1mg/mL
Cross Reactivity Virus
Antigen EBV Early Antigens
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Conjugate Unlabeled