Please login to your online account to display your discounted pricing

Mouse anti-EBV Early Antigens, Clone: 1108-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451601

  • / Each
 

Description & Specifications

Specifications

Antigen EBV Early Antigens
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone 1108-1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Epstein-Barr virus
Reconstitution Protein A purified
Target Species Virus