Please login to your online account to display your discounted pricing

Mouse anti-E-Selectin/CD62E, Clone: P2H3, Novus Biologicals™

$230.00 - $285.00

Specifications

Antigen E-Selectin/CD62E
Clone P2H3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD62E.
Gene ID (Entrez) 6401
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14339301

 
novus biologicals canada
NBP14339301MG
0.1mg Each for $285.00

NBP14339303

 
novus biologicals canada
NBP1433930025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen E-Selectin/CD62E
Clone P2H3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD62E.
Gene ID (Entrez) 6401
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation