Please login to your online account to display your discounted pricing

Mouse anti-Dystrophin, Clone: SPM499, Novus Biologicals™

$307.00 - $518.93

Specifications

Antigen Dystrophin
Clone SPM499
Immunogen Synthetic peptide corresponding to C-terminus of human dystrophin.
Gene ID (Entrez) 1756
Formulation 10mM PBS/1% BSA buffer pH 7.4. Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13578401

 
novus biologicals canada
NBP13578401ML
0.1mL Each for $307.00

NBP13578405

 
novus biologicals canada
NBP13578405ML
0.5mL Each for $363.90

NBP13578410

 
novus biologicals canada
NBP13578410ML
1mL Each for $518.93
Description & Specifications

Specifications

Antigen Dystrophin
Clone SPM499
Immunogen Synthetic peptide corresponding to C-terminus of human dystrophin.
Gene ID (Entrez) 1756
Formulation 10mM PBS/1% BSA buffer pH 7.4. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)