Please login to your online account to display your discounted pricing

Mouse anti-Dystrophin, Clone: SPM499, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13578410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13578410

  • / Each
 

Description & Specifications

Specifications

Antigen Dystrophin
Format Affinity Purified
Host Species Mouse
Primary or Secondary Primary
Gene ID (Entrez) 1756
Cross Reactivity Human
Cross Reactivity Rat
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide corresponding to C-terminus of human dystrophin.
Isotype IgG1, κ
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone SPM499
Conjugate Unlabeled
Formulation 10mM PBS/1% BSA buffer pH 7.4. Immunogen affinity purified.
Purification Method Affinity Purified
Test Specificity This product is specific for Dystrophin