Please login to your online account to display your discounted pricing

Mouse anti-Dystrophin, Clone: SPM499, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13578405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13578405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity This product is specific for Dystrophin
Formulation 10mM PBS/1% BSA buffer pH 7.4. Immunogen affinity purified.
Immunogen Synthetic peptide corresponding to C-terminus of human dystrophin.
Host Species Mouse
Isotype IgG1, κ
Format Affinity Purified
Antigen Dystrophin
Gene ID (Entrez) 1756
Storage Requirements Store at 4°C. Do not freeze.
Clone SPM499
Cross Reactivity Human
Cross Reactivity Rat
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO