Please login to your online account to display your discounted pricing

Mouse anti-Dystrophin, Clone: SPM499, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13578470ML

 View all options for this product


For Research Use Only

Catalog No. NBP13578470


Description & Specifications


Antigen Dystrophin
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SPM499
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Affinity Purified
Formulation 10mM PBS/1% BSA buffer pH 7.4. Immunogen affinity purified.
Host Species Mouse
Immunogen Synthetic peptide corresponding to C-terminus of human dystrophin.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 7mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1756
Test Specificity This product is specific for Dystrophin