Please login to your online account to display your discounted pricing

Mouse anti-DPPIV/CD26, Clone: 2A6, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen DPPIV/CD26
Clone 2A6
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD26.
Gene ID (Entrez) 1803
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14342901

 
novus biologicals canada
NBP14342901MG
0.1mg Each for $285.00

NBP14342903

 
novus biologicals canada
NBP1434290025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen DPPIV/CD26
Clone 2A6
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD26.
Gene ID (Entrez) 1803
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry