Please login to your online account to display your discounted pricing

Mouse anti-DPPIV/CD26, Clone: 2A6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434290025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14342903

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Conjugate Unlabeled
Immunogen The immunogen for this antibody was CD26.
Quantity 0.025mg
Antigen DPPIV/CD26
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone 2A6
Concentration 0.5mg/mL
Isotype IgG1, κ
Format Affinity Purified
Applications Flow Cytometry
Purification Method Affinity Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1803
Cross Reactivity Human