Please login to your online account to display your discounted pricing

Mouse anti-DPPIV/CD26, Clone: 2A6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14342901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14342901

  • / Each
 

Description & Specifications

Specifications

Antigen DPPIV/CD26
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Gene ID (Entrez) 1803
Host Species Mouse
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Immunogen The immunogen for this antibody was CD26.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone 2A6
Cross Reactivity Human