Please login to your online account to display your discounted pricing

Mouse anti-DOG1/TMEM16A, Clone: SPM580, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23284110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23284110

  • / Each
 

Description & Specifications

Specifications

Antigen DOG1/TMEM16A
Format Purified
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 1.0mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 55107
Conjugate Unlabeled
Gene Accession No. Q5XXA6
Host Species Mouse
Immunogen Recombinant human DOG-1 protein
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone SPM580
Concentration 0.2mg/mL
Cross Reactivity Human