Please login to your online account to display your discounted pricing

Mouse anti-DOG1/TMEM16A, Clone: SPM580, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23284101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23284101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen DOG1/TMEM16A
Formulation PBS with 0.05% BSA.
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Gene Accession No. Q5XXA6
Immunogen Recombinant human DOG-1 protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 55107
Reconstitution Protein A purified
Clone SPM580
Concentration 0.2mg/mL