Please login to your online account to display your discounted pricing

Mouse anti-DOG1/TMEM16A, Clone: DG1/447 + DOG-1.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23428501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23428501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Gene Accession No. Q5XXA6
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human DOG-1 protein (DG1/447); A synthetic peptide from human DOG-1 protein (MSDFVDWVIPDIPKDISQQIHK-EKVLMVELFMREEQDKQQLLETCMEKER QKDEPPCNHHNTKACPDSLGSPAPSHAYHGGVL)
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 55107
Cross Reactivity Human
Antigen DOG1/TMEM16A
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone DG1/447 + DOG-1.1
Concentration 0.2mg/mL