Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV101, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433805

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Host Species Mouse
Immunogen Recombinant CMV-p65 protein
Regulatory Status RUO
Format Purified
Quantity 0.5mL
Test Specificity Cytomegalovirus
Formulation PBS with 0.05% BSA.
Clone CMV101
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Primary or Secondary Primary
Concentration 0.2mg/mL
Isotype IgG1, κ
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Antigen Cytomegalovirus p65
Reconstitution Protein A purified
Cross Reactivity Virus