Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV101, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459601

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Regulatory Status RUO
Conjugate Unlabeled
Cross Reactivity Virus
Antigen Cytomegalovirus p65
Format Purified
Immunogen Recombinant CMV-p65 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Cytomegalovirus
Reconstitution Protein A purified
Clone CMV101
Concentration 1mg/mL