Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV100, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytomegalovirus p65
Clone CMV100
Concentration 0.2mg/mL
Immunogen Recombinant CMV-p65 protein
Formulation PBS with 0.05% BSA.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23433205

 
novus biologicals canada
NBP23433205ML
0.5mL Each for $321.00

NBP23433201

 
novus biologicals canada
NBP23433201ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen Cytomegalovirus p65
Clone CMV100
Concentration 0.2mg/mL
Immunogen Recombinant CMV-p65 protein
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Virus
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence