Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV100, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Immunogen Recombinant CMV-p65 protein
Format Purified
Cross Reactivity Virus
Regulatory Status RUO
Conjugate Unlabeled
Host Species Mouse
Primary or Secondary Primary
Isotype IgG1, κ
Purification Method Purified
Test Specificity Cytomegalovirus
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Antigen Cytomegalovirus p65
Quantity 0.1mL
Clone CMV100
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.