Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV100, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433205

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Virus
Host Species Mouse
Format Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Clone CMV100
Purification Method Purified
Immunogen Recombinant CMV-p65 protein
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Antigen Cytomegalovirus p65
Concentration 0.2mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Cytomegalovirus
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Isotype IgG1, κ
Regulatory Status RUO
Quantity 0.5mL