missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV100, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytomegalovirus p65
Clone CMV100
Concentration 1mg/mL
Immunogen Recombinant CMV-p65 protein
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23459501 Novus Biologicals Canada
NBP23459501MG
0.1mg Each for $384.00
NBP23459502 Novus Biologicals Canada
NBP23459502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Cytomegalovirus p65
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
CMV100
Recombinant CMV-p65 protein
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Immunocytochemistry,Immunofluorescence