Please login to your online account to display your discounted pricing

Mouse anti-Cytomegalovirus p65, Clone: CMV100, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459501

  • / Each
 

Description & Specifications

Specifications

Antigen Cytomegalovirus p65
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Cross Reactivity Virus
Host Species Mouse
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Immunogen Recombinant CMV-p65 protein
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Cytomegalovirus
Clone CMV100
Concentration 1mg/mL
Conjugate Unlabeled