Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, pan, Clone: SPM115 + SPM116, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434705

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Antigen Cytokeratin, pan
Host Species Mouse
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which include CK1, CK3, CK4, CK5, CK6, CK8, CK10, CK14, CK15, CK16, and CK19. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Immunogen Human epidermal keratin
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Clone SPM115 + SPM116
Concentration 0.2mg/mL
Conjugate Unlabeled