Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, pan, Clone: SPM115 + SPM116, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434701

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Immunogen Human epidermal keratin
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Primary or Secondary Primary
Quantity 0.1mL
Concentration 0.2mg/mL
Test Specificity This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which include CK1, CK3, CK4, CK5, CK6, CK8, CK10, CK14, CK15, CK16, and CK19. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.
Antigen Cytokeratin, pan
Isotype IgG1
Format Purified
Purification Method Purified
Host Species Mouse
Formulation PBS with 0.05% BSA. Protein G purified.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM115 + SPM116