Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, pan, Clone: SPM115 + SPM116, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytokeratin, pan
Clone SPM115 + SPM116
Concentration 1mg/mL
Immunogen Human epidermal keratin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23439401

 
novus biologicals canada
NBP23439401MG
0.1mg Each for $384.00

NBP23439402

 
novus biologicals canada
NBP23439402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cytokeratin, pan
Clone SPM115 + SPM116
Concentration 1mg/mL
Immunogen Human epidermal keratin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting