Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, pan, Clone: SPM115 + SPM116, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Quantity 0.1mg
Conjugate Unlabeled
Antigen Cytokeratin, pan
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Human epidermal keratin
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which include CK1, CK3, CK4, CK5, CK6, CK8, CK10, CK14, CK15, CK16, and CK19. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.
Reconstitution Protein A purified
Clone SPM115 + SPM116
Concentration 1mg/mL