Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, HMW, Clone: 34BE12, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin, HMW
Clone 34BE12
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Gene Accession No. P04264
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP22941701

 
novus biologicals canada
NBP22941701ML
0.1mL Each for $139.00

NBP23434001

 
novus biologicals canada
NBP23434001ML
0.1mL Each for $139.00

NBP23434005

 
novus biologicals canada
NBP23434005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin, HMW
Clone 34BE12
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Gene Accession No. P04264
Gene ID (Entrez) 51350
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting