Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, HMW, Clone: 34BE12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434005

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Isotype IgG1, κ
Primary or Secondary Primary
Immunogen Solubilized keratin extract from human stratum corneum
Gene Accession No. P04264
Gene ID (Entrez) 51350
Concentration 0.2mg/mL
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.5mL
Regulatory Status RUO
Reconstitution Protein A purified
Formulation PBS with 0.05% BSA.
Purification Method Purified
Clone 34BE12
Antigen Cytokeratin, HMW