Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, HMW, Clone: 34BE12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Primary or Secondary Primary
Antigen Cytokeratin, HMW
Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Solubilized keratin extract from human stratum corneum
Quantity 0.1mL
Regulatory Status RUO
Gene ID (Entrez) 51350
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. P04264
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone 34BE12
Concentration 0.2mg/mL