Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, HMW, Clone: 34BE12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1, κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Primary or Secondary Primary
Gene Accession No. P04264
Gene ID (Entrez) 51350
Host Species Mouse
Quantity 0.1mg
Immunogen Solubilized keratin extract from human stratum corneum
Purification Method Purified
Regulatory Status RUO
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Antigen Cytokeratin, HMW
Clone 34BE12
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.