Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Basic (Type II or HMW), Clone: SPM591, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin, Basic (Type II or HMW)
Clone SPM591
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Formulation PBS with 0.05% BSA.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23438801

 
novus biologicals canada
NBP23438801ML
0.1mL Each for $139.00

NBP23438805

 
novus biologicals canada
NBP23438805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin, Basic (Type II or HMW)
Clone SPM591
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting