Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Basic (Type II or HMW), Clone: SPM591, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438801

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin, Basic (Type II or HMW)
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM591
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Solubilized keratin extract from human stratum corneum
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Test Specificity This antibody recognizes CK1, CK5, CK10 and CK14. This MAb shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this antibody.
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled