missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Basic (Type II or HMW), Clone: SPM591, Azide Free, Novus Biologicals™

$396.83 - $676.00

Specifications

Antigen Cytokeratin, Basic (Type II or HMW)
Clone SPM591
Concentration 1mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23443501 Novus Biologicals Canada
NBP23443501MG
0.1mg Each for $396.83
NBP23443502 Novus Biologicals Canada
NBP23443502MG
0.2mg Each for $676.00
Specifications Description & Specifications

Specifications

Cytokeratin, Basic (Type II or HMW)
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM591
Solubilized keratin extract from human stratum corneum
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Western Blot