Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Basic (Type II or HMW), Clone: SPM591, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443502

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Reconstitution Protein A purified
Antigen Cytokeratin, Basic (Type II or HMW)
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Regulatory Status RUO
Immunogen Solubilized keratin extract from human stratum corneum
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Quantity 0.2mg
Purification Method Purified
Primary or Secondary Primary
Test Specificity This antibody recognizes CK1, CK5, CK10 and CK14. This MAb shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this antibody.
Isotype IgG1, κ
Clone SPM591