Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Basic (Type II or HMW), Clone: SPM116, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434805

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Immunogen Solubilized keratin extract from human stratum corneum
Formulation PBS with 0.05% BSA. Protein G purified.
Regulatory Status RUO
Conjugate Unlabeled
Antigen Cytokeratin, Basic (Type II or HMW)
Concentration 0.2mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Isotype IgG1
Clone SPM116
Test Specificity This MAb recognizes basic (Type II or HMW) cytokeratins, which include 67kDa (CK1); 64kDa (CK3); 59kDa (CK4); 58kDa (CK5); 56kDa (CK6); 52kDa (CK8). This MAb recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily.
Purification Method Purified
Host Species Mouse